missing translation for 'onlineSavingsMsg'
Learn More

Aminopterin Hydrate 97.0+%, TCI America™

Catalog No. p-7116882
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A041025MG 25 mg
A0410100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A041025MG Supplier TCI America Supplier No. A041025MG

Chemical Identifiers

CAS 54-62-6
Molecular Formula C19H20N8O5
Molecular Weight (g/mol) 440.42
MDL Number MFCD00036692
InChI Key TVZGACDUOSZQKY-LBPRGKRZSA-N
Synonym 4-Aminofolic Acid, 4-Amino-PGA, 4-Aminopteroyl-L-glutamic Acid
PubChem CID 169371
ChEBI CHEBI:22526
IUPAC Name (2S)-2-[[4-[(2,4-diaminopteridin-6-yl)methylamino]benzoyl]amino]pentanedioic acid
SMILES C1=CC(=CC=C1C(=O)NC(CCC(=O)O)C(=O)O)NCC2=CN=C3C(=N2)C(=NC(=N3)N)N

Specifications

Melting Point 222°C
Color Yellow
Quantity 25 mg
UN Number 2811
Formula Weight 440.42
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Aminopterin Hydrate
RTECSNumber MA1050000
TSCA Yes
Recommended Storage Refrigerator