missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Aminopeptidase N (CD13), Human Serum

Catalog No. 16460510UG
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16460510UG 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16460510UG Supplier MilliporeSigma™ Supplier No. 16460510UG

Prepared from human serum that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV

  • Native aminopeptidase N purified from human serum
  • Aminopeptidase N is a single-chain, integral membrane protease with broad specificity that functions to remove N-terminal amino acids from peptides
  • It also acts as a receptor for coronavirus and is known to play a role in angiogenesis, HIV entry, and in degradation of extracellular matrix as a result of tumor invasion

Specifications

Formulation In 150mM NaCl, 50mM Tris-HCl, 1mM CaCl₂, pH 8.0
Purity ≥92% by SDS-PAGE
Content And Storage ≤-70°C
Form Liquid
Quantity 10 μg
Synonym APN/CD13, Aminopeptidase N
Product Type Aminopeptidase