missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Aminopeptidase M, Porcine Kidney

Catalog No. 16459850U
Change view
Click to view available options
Quantity:
50 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16459850U 50 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16459850U Supplier MilliporeSigma™ Supplier No. 16459850U

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Useful in peptide sequencing

  • Native amino peptidase M from porcine kidney
  • Metalloprotease that hydrolyzes N-terminal amino acids from almost all unsubstituted oligopeptides
  • Does not cleave X-Pro bonds or N-blocked amino acids
  • Inhibited by EDTA, bestatin, and amastatin

Specifications

Concentration Please refer to vial label for lot-specific concentration
Formulation Crystalline suspension in 3.2 M (NH₄)₂SO₄, 10 mM MgCl₂, 10 mM Tris-HCl, pH 7.5.
pH Range Optimum 7.0 to 7.5
CAS 9054-63-1
Content And Storage 2°C to 8°C
Form Crystalline Suspension
Quantity 50 U
Synonym Aminopeptidase M
Solubility Physiological Buffers, Water
Product Type Aminopeptidase