missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aminoacetamidine Dihydrobromide 95.0+%, TCI America™
$229.29 - $788.49
Chemical Identifiers
| CAS | 69816-37-1 |
|---|---|
| Molecular Formula | C2H9Br2N3 |
| Molecular Weight (g/mol) | 234.923 |
| MDL Number | MFCD00671486 |
| InChI Key | SLCZNGZFOVAAED-UHFFFAOYSA-N |
| PubChem CID | 16211382 |
| IUPAC Name | 2-aminoethanimidamide;dihydrobromide |
| SMILES | C(C(=N)N)N.Br.Br |
Chemical Identifiers
| 69816-37-1 | |
| 234.923 | |
| SLCZNGZFOVAAED-UHFFFAOYSA-N | |
| 2-aminoethanimidamide;dihydrobromide |
| C2H9Br2N3 | |
| MFCD00671486 | |
| 16211382 | |
| C(C(=N)N)N.Br.Br |
Specifications
| 69816-37-1 | |
| C2H9Br2N3 | |
| 1 g | |
| C(C(=N)N)N.Br.Br | |
| 234.923 | |
| 234.92 | |
| Crystalline Powder |
| White-Yellow | |
| MFCD00671486 | |
| SLCZNGZFOVAAED-UHFFFAOYSA-N | |
| 2-aminoethanimidamide;dihydrobromide | |
| 16211382 | |
| ≥95.0% (T) | |
| Aminoacetamidine Dihydrobromide |
Safety and Handling
TSCA : No