missing translation for 'onlineSavingsMsg'
Learn More

Aminoacetamidine Dihydrobromide 95.0+%, TCI America™

Catalog No. a13615g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
A13615G 5 g
A13611G 1 g
2 options

Catalog No. A13615G

Supplier: TCI America A13615G

Only null left

Chemical Identifiers

CAS 69816-37-1
Molecular Formula C2H9Br2N3
Molecular Weight (g/mol) 234.923
MDL Number MFCD00671486
InChI Key SLCZNGZFOVAAED-UHFFFAOYSA-N
PubChem CID 16211382
IUPAC Name 2-aminoethanimidamide;dihydrobromide
SMILES C(C(=N)N)N.Br.Br

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 234.92
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Aminoacetamidine Dihydrobromide