missing translation for 'onlineSavingsMsg'
Learn More

Amino Acid Standard, Analytical Standard, MilliporeSigma™ Supelco™

Amino acids in this standard are 2.5 mumoles per mL in 0.1 N HCl, except L-cystine at 1.25 mumoles per mL.

$181.52 - $814.29

Specifications

Description The following components are included in the amino acid standard:. L-Alanine. Ammonium chloride. L-Arginine. L-Aspartic acid. L-Cystine. L-Glutamic acid. Glycine. L-Histidine. L-Isoleucine. L-Leucine. L-Lysine. L-Methionine. L-Phenylalanine. L-Proline. L-Serine. L-Threonine. L-Tyrosine. L-Valine
Packaging Glass Bottle
Physical Form Neat
Grade Analytical Standard
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111004841
View Documents
Merck Emd Millipore
AAS185ML
5 mL
Each for $181.52
Only null left
 
111004483
View Documents
Merck Emd Millipore
AAS1810X1ML
10 x 1 mL
Each for $337.26
Only null left
 
111004546
View Documents
Merck Emd Millipore
AAS1810ML
10 mL
Each for $308.73
Only null left
 
111003908
View Documents
Merck Emd Millipore
AAS185X5ML
5 x 5 mL
Each for $814.29
Only null left
 
Description

Description

Specifications

Specifications

The following components are included in the amino acid standard:. L-Alanine. Ammonium chloride. L-Arginine. L-Aspartic acid. L-Cystine. L-Glutamic acid. Glycine. L-Histidine. L-Isoleucine. L-Leucine. L-Lysine. L-Methionine. L-Phenylalanine. L-Proline. L-Serine. L-Threonine. L-Tyrosine. L-Valine
Glass Bottle
Neat
Analytical Standard
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'