missing translation for 'onlineSavingsMsg'
Learn More

Amino Acid Standard, Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111004841
Change view
Click to view available options
Quantity:
5 mL
10 mL
5 x 5 mL
10 x 1 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111004841 5 mL
111004483 10 x 1 mL
111004546 10 mL
111003908 5 x 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 111004841 Supplier Merck Emd Millipore Supplier No. AAS185ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Amino acids in this standard are 2.5 mumoles per mL in 0.1 N HCl, except L-cystine at 1.25 mumoles per mL.

Specifications

Description The following components are included in the amino acid standard:. L-Alanine. Ammonium chloride. L-Arginine. L-Aspartic acid. L-Cystine. L-Glutamic acid. Glycine. L-Histidine. L-Isoleucine. L-Leucine. L-Lysine. L-Methionine. L-Phenylalanine. L-Proline. L-Serine. L-Threonine. L-Tyrosine. L-Valine
Packaging Glass Bottle
Physical Form Neat
Quantity 5 mL
Grade Analytical Standard