missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amine-Sulfuric Acid Reagent, for Sulfide Determination (Methylene Blue Method), Ricca Chemical
Sulfide Determination (Methylene Blue Method)
$257.84 - $305.97
Specifications
| Chemical Name or Material | Amine-Sulfuric Acid Reagent |
|---|---|
| Color | Tan to Yellow |
| CAS | 7664-93-9,7664-93-9,62778-12-5 |
| CAS Min % | 51.0,46.79,0.34 |
| CAS Max % | 53.0,48.7,0.36 |
Specifications
| Amine-Sulfuric Acid Reagent | |
| 7664-93-9,7664-93-9,62778-12-5 | |
| 53.0,48.7,0.36 | |
| Safety-coated Glass Bottle | |
| Liquid |
| Tan to Yellow | |
| 51.0,46.79,0.34 | |
| Laboratory | |
| <3 |