missing translation for 'onlineSavingsMsg'
Learn More

Amine-Sulfuric Acid Reagent, for Sulfide Determination (Methylene Blue Method), Ricca Chemical

Catalog No. 61016
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61016 500 mL
61032 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 61016 Supplier Ricca Chemical Company Supplier No. 61016

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sulfide Determination (Methylene Blue Method)

Specifications

Chemical Name or Material Amine-Sulfuric Acid Reagent
Color Tan to Yellow
CAS 7664-93-9, 62778-12-5
CAS Min % 51.0, 46.79, 0.34
CAS Max % 53.0, 48.7, 0.36
Grade Laboratory
Packaging Safety-coated Glass Bottle
pH <3
Quantity 500 mL
Physical Form Liquid