Learn More
Amikacin disulfate
An aminoglycoside antibiotic derived from Kanamycin A | CAS: 39831-55-5 | C22H47N5O21S2 | 781.75 g/mol
$287.26 - $797.36
Chemical Identifiers
| CAS | 39831-55-5 |
|---|---|
| Molecular Formula | C22H47N5O21S2 |
| Molecular Weight (g/mol) | 781.75 |
| MDL Number | MFCD00167475,MFCD00167475 |
| InChI Key | FXKSEJFHKVNEFI-GCZBSULCSA-N |
| PubChem CID | 134129615 |
| IUPAC Name | (2S)-4-amino-N-[(1R,2S,3S,4R,5S)-5-amino-2-{[(2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-4-{[(2R,3R,4S,5S,6R)-6-(aminomethyl)-3,4,5-trihydroxyoxan-2-yl]oxy}-3-hydroxycyclohexyl]-2-hydroxybutanamide; bis(sulfuric acid) |
| SMILES | OS(O)(=O)=O.OS(O)(=O)=O.NCC[C@H](O)C(=O)N[C@@H]1C[C@H](N)[C@@H](O[C@H]2O[C@H](CN)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@H]1O[C@H]1O[C@H](CO)[C@@H](O)[C@H](N)[C@H]1O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ6386214
|
Thermo Scientific Chemicals
J6386214 |
25 g |
Each for $287.26
|
|
|||||
|
AAJ6386222
|
Thermo Scientific Chemicals
J6386222 |
100 g |
Each for $797.36
|
|
|||||
Description
Amikacin disulfate is used as an aminoglycoside antibiotic derived from Kanamycin A, used for treating infections with multidrug resistant Gram negative bacteria such as Pseudomonas aeruginosa, Acinetobacter, and Enterobacter. It is commonly used in the treatment of drug-resistant mycobacteria. It is used to study organism-directed delivery of antibiotics as well as drug resistance.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAmikacin disulfate is used as an aminoglycoside antibiotic derived from Kanamycin A, used for treating infections with multidrug resistant Gram negative bacteria such as Pseudomonas aeruginosa, Acinetobacter, and Enterobacter. It is commonly used in the treatment of drug-resistant mycobacteria. It is used to study organism-directed delivery of antibiotics as well as drug resistance.
Solubility
Soluble in water at 50mg/ml with warming
Notes
Potency: 674-786 micrograms per mg
Chemical Identifiers
| 39831-55-5 | |
| 781.75 | |
| FXKSEJFHKVNEFI-GCZBSULCSA-N | |
| (2S)-4-amino-N-[(1R,2S,3S,4R,5S)-5-amino-2-{[(2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-4-{[(2R,3R,4S,5S,6R)-6-(aminomethyl)-3,4,5-trihydroxyoxan-2-yl]oxy}-3-hydroxycyclohexyl]-2-hydroxybutanamide; bis(sulfuric acid) |
| C22H47N5O21S2 | |
| MFCD00167475,MFCD00167475 | |
| 134129615 | |
| OS(O)(=O)=O.OS(O)(=O)=O.NCC[C@H](O)C(=O)N[C@@H]1C[C@H](N)[C@@H](O[C@H]2O[C@H](CN)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@H]1O[C@H]1O[C@H](CO)[C@@H](O)[C@H](N)[C@H]1O |
Specifications
| Amikacin Disulfate | |
| 39831-55-5 | |
| White | |
| C22H47N5O21S2 | |
| 6172633 | |
| Soluble in water at 50mg/ml with warming | |
| OS(O)(=O)=O.OS(O)(=O)=O.NCC[C@H](O)C(=O)N[C@@H]1C[C@H](N)[C@@H](O[C@H]2O[C@H](CN)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@H]1O[C@H]1O[C@H](CO)[C@@H](O)[C@H](N)[C@H]1O | |
| 781.75 | |
| 781.77 |
| Powder | |
| 220°C to 230°C | |
| 25 g | |
| MFCD00167475,MFCD00167475 | |
| 14,405 | |
| FXKSEJFHKVNEFI-GCZBSULCSA-N | |
| (2S)-4-amino-N-[(1R,2S,3S,4R,5S)-5-amino-2-{[(2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-4-{[(2R,3R,4S,5S,6R)-6-(aminomethyl)-3,4,5-trihydroxyoxan-2-yl]oxy}-3-hydroxycyclohexyl]-2-hydroxybutanamide; bis(sulfuric acid) | |
| 134129615 | |
| Amikacin disulfate |
Safety and Handling
EINECSNumber : 254-648-6
RTECSNumber : WK1961200
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only