missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Amicon™ Ultra-15 Centrifugal Filter Units

Catalog No. UFC900308
Change view
Click to view available options
MWCO:
10 kDa
100 kDa
3 kDa
30 kDa
50 kDa
Quantity:
24Pack
8Pack
96 Pack
96Pack
Pack of 96
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. MWCO Quantity
UFC900308 3 kDa 8Pack
UFC900396 3 kDa 96 Pack
UFC901008 10 kDa 8Pack
UFC901024 10 kDa 24Pack
UFC901096 10 kDa Pack of 96
UFC903008 30 kDa 8Pack
UFC903024 30 kDa 24Pack
UFC903096 30 kDa 96Pack
UFC905008 50 kDa 8Pack
UFC905096 50 kDa 96Pack
UFC910024 100 kDa 24Pack
UFC910096 100 kDa 96Pack
UFC900324 3 kDa 24Pack
UFC910008 100 kDa 8Pack
UFC905024 50 kDa 24Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Catalog No. UFC900308 Supplier MilliporeSigma™ Supplier No. UFC900308

Achieve both high sample recovery and fast sample processing. EMD Amicon™ Ultra Centrifugal Filter Units feature regenerated cellulose membranes that are ideal for protein & nucleic acid purification, concentration and desalting.

  • High-recovery Ultracel™-PL regenerated cellulose membranes with MWCOs of 3, 10, 30, 50 or 100kDa
  • Vertical membrane reduces polarization for ultrafast spin times (as little as 10 to 15 min.)
  • Heat-sealed membrane eliminates possibility of downstream extractables
  • Universal rotor compatibility
  • High retentate recovery of >90%
  • High concentration factors of 80- to 100-fold (minimum final concentrate volume: 200μL)
  • Easy sample monitoring due to translucent housing and volume gradations
  • Direct pipetter sample access eliminates processing step to recover concentrate
Centrifugal filter enables
  • Concentration of biological samples containing antigens, antibodies, enzymes, nucleic acids, or microorganisms
  • Purification of macromolecular components found in tissue culture extracts or cell lysates
  • Concentration of dilute or prepurified proteins from column eluents
  • Desalting and buffer exchange

Specifications

Disposable Single-use
Material (Membrane) High-recovery Ultracel-PL Regenerated Cellulose
MWCO 3 kDa
Sterility Non-sterile
Filtration Area 7.6 cm2
For Use With (Application) Protein purification, protein concentration and desalting
Housing Material Regenerated Cellulose (RC)
Includes Two microcentrifuge tubes
Quantity 8Pack
Type Centrifugal Filters
Volume (Metric) 15 mL
Length (Metric) 121 mm
Diameter (Metric) 29.7 mm
Show More Show Less