missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Amicon™ Ultra-0.5 Centrifugal Filter Units
Description
Includes
Two microcentrifuge tubes
- High-recovery Ultracel™-PL regenerated cellulose membranes with MWCOs of 3, 10, 30, 50 or 100kDa
- Vertical membrane reduces polarization for ultrafast spin times (as little as 10 to 15 min.)
- Heat-sealed membrane eliminates possibility of downstream extractables
- Universal rotor compatibility
- High retentate recovery of >90%
- High concentration factors of 80- to 100-fold (minimum final concentrate volume: 200μL)
- Easy sample monitoring due to translucent housing and volume gradations
- Direct pipetter sample access eliminates processing step to recover concentrate
- Concentration of biological samples containing antigens, antibodies, enzymes, nucleic acids, or microorganisms
- Purification of macromolecular components found in tissue culture extracts or cell lysates
- Concentration of dilute or prepurified proteins from column eluents
- Desalting and buffer exchange
Specifications
Specifications
| Disposable | Single-use |
| Material (Membrane) | High-recovery Ultracel-PL Regenerated Cellulose |
| MWCO | 100 kDa |
| Sterility | Non-sterile |
| Filtration Area | 1 cm2 |
| For Use With (Application) | Protein purification, protein concentration and desalting |
| Housing Material | Regenerated Cellulose (RC) |
| Includes | Two microcentrifuge tubes |
| Max. Force | 14,000 xG for concentration spin; 1000 xG for reverse spin |
| Quantity | 24Pack |
| Show More |