missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Amicon™ Ultra-0.5 Centrifugal Filter Units

Catalog No. UFC500324
Change view
Click to view available options
MWCO:
10 kDa
100 kDa
3 kDa
30 kDa
50 kDa
Quantity:
24Pack
500Pack
96 Pack
96Pack
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. MWCO Quantity
UFC500324 3 kDa 24Pack
UFC500396 3 kDa 96Pack
UFC5003BK 3 kDa 500Pack
UFC501024 10 kDa 24Pack
UFC501096 10 kDa 96 Pack
UFC5010BK 10 kDa 500Pack
UFC503024 30 kDa 24Pack
UFC503096 30 kDa 96Pack
UFC5030BK 30 kDa 500Pack
UFC505024 50 kDa 24Pack
UFC505096 50 kDa 96Pack
UFC5050BK 50 kDa 500Pack
UFC510024 100 kDa 24Pack
UFC510096 100 kDa 96Pack
UFC5100BK 100 kDa 500Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Catalog No. UFC500324 Supplier MilliporeSigma™ Supplier No. UFC500324
Edge

Includes

Two microcentrifuge tubes

Achieve both high sample recovery and fast sample processing. EMD Amicon™ Ultra Centrifugal Filter Units feature regenerated cellulose membranes that are ideal for protein & nucleic acid purification, concentration and desalting.

  • High-recovery Ultracel™-PL regenerated cellulose membranes with MWCOs of 3, 10, 30, 50 or 100kDa
  • Vertical membrane reduces polarization for ultrafast spin times (as little as 10 to 15 min.)
  • Heat-sealed membrane eliminates possibility of downstream extractables
  • Universal rotor compatibility
  • High retentate recovery of >90%
  • High concentration factors of 80- to 100-fold (minimum final concentrate volume: 200μL)
  • Easy sample monitoring due to translucent housing and volume gradations
  • Direct pipetter sample access eliminates processing step to recover concentrate
Centrifugal filter enables
  • Concentration of biological samples containing antigens, antibodies, enzymes, nucleic acids, or microorganisms
  • Purification of macromolecular components found in tissue culture extracts or cell lysates
  • Concentration of dilute or prepurified proteins from column eluents
  • Desalting and buffer exchange

Specifications

Disposable Single-use
Material (Membrane) High-recovery Ultracel-PL Regenerated Cellulose
MWCO 3 kDa
Sterility Non-sterile
Filtration Area 1 cm2
For Use With (Application) Protein purification, protein concentration and desalting
Housing Material Regenerated Cellulose (RC)
Includes Two microcentrifuge tubes
Max. Force 14,000 xG for concentration spin; 1000 xG for reverse spin
Quantity 24Pack
Type Centrifugal Filters
Volume (Metric) 0.5 mL
Length (English) 0.196 in.
Length (Metric) 4.99 cm
Diameter (Metric) 10.8 mm
Show More Show Less