missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ AMG 18 hydrochloride
Description
AMG 18 hydrochloride is a potent and selective IRE1α inhibitor (IC50 values are 13 and 99 nM for recombinant and cellular IRE1α, respectively). Exhibits selectivity for IRE1α in a panel of over 100 other kinases.
Specifications
Specifications
| CAS | 2250019-92-0 |
| Synonym | 2-Chloro-N-[6-methyl-5-[[3-[2-[(3S)-3-piperidinylamino]-4-pyrimidinyl]-2-pyridinyl]oxy]-1-naphthalenyl]benzenesulfonamide hydrochloride |
| Quantity | 5 mg |
| Product Line | Tocris |
| Molecular Formula | C31H29ClN6O3S.HCl |
| Formula Weight | 637.58 |
| Purity | 98% |