missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Amersham™ Streptavidin-HRP Conjugate
Streptavidin conjugated to reporter enzyme molecules for use in blotting and in situ hybridization applications
$132.02 - $404.15
Specifications
| For Use With (Application) | Suitable for use in ECL™, western blotting ,ELISA, immunocytochemistry |
|---|---|
| Includes | 2mL of Phosphate Buffered Saline (0.1M Sodium Phosphate, 0.1M Sodium Chloride, pH 7.4) containing 1%(w/v) Bovine Serum Albumin and an antimicrobial agent |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
45-000-765
|
Cytiva
RPN12312ML |
2 mL |
Each for $404.15
|
|
|||||
|
45-001-277
|
Cytiva
RPN1231100UL |
100 mL |
Each for $132.02
|
|
|||||
Description
- High specificity reagents
- Conjugation achieved using the periodate coupling method described by Nakane and co-workers
- Sold on the basis of titer in an ELISA and performance in a western blot
Specifications
| Suitable for use in ECL™, western blotting ,ELISA, immunocytochemistry | |
| 2mL of Phosphate Buffered Saline (0.1M Sodium Phosphate, 0.1M Sodium Chloride, pH 7.4) containing 1%(w/v) Bovine Serum Albumin and an antimicrobial agent |