Learn More
Ambroxol hydrochloride, 98%
CAS: 23828-92-4 | C13H18Br2N2O | 378.108 g/mol
$69.01 - $255.97
Chemical Identifiers
| CAS | 23828-92-4 |
|---|---|
| Molecular Formula | C13H18Br2N2O |
| Molecular Weight (g/mol) | 378.108 |
| MDL Number | MFCD00078932 |
| InChI Key | JBDGDEWWOUBZPM-UHFFFAOYSA-N |
| Synonym | ambroxol, ambroxolum, bisolvon metabolite viii, bromhexine-metabolite viii, 4-2-amino-3,5-dibromobenzyl amino cyclohexanol, ambroxol inn:ban, ambroxol ep impurity d, ambroxol base, ambroxolum inn-latin, rac-cis-ambroxol |
| PubChem CID | 2132 |
| IUPAC Name | 4-[(2-amino-3,5-dibromophenyl)methylamino]cyclohexan-1-ol |
| SMILES | C1CC(CCC1NCC2=CC(=CC(=C2N)Br)Br)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ6371203
|
Thermo Scientific Chemicals
J6371203 |
1 g |
Each for $69.01
|
|
|||||
|
AAJ6371206
|
Thermo Scientific Chemicals
J6371206 |
5 g |
Each for $255.97
|
|
|||||
Description
Ambroxol hydrochloride is a medication indicated to alleviate chest congestion associated with conditions that include bronchitis, pneumonia, bronchospasm asthma, cough, and allergy. Preclinically, ambroxol, the active ingredient of Mucosolvan, has been shown to increase respiratory tract secretion. It enhances pulmonary surfactant production and stimulates ciliary activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAmbroxol hydrochloride is a medication indicated to alleviate chest congestion associated with conditions that include bronchitis, pneumonia, bronchospasm asthma, cough, and allergy. Preclinically, ambroxol, the active ingredient of Mucosolvan, has been shown to increase respiratory tract secretion. It enhances pulmonary surfactant production and stimulates ciliary activity.
Solubility
Slightly soluble in water (∼5 mg/ml) or ethanol (∼5 mg/ml).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Chemical Identifiers
| 23828-92-4 | |
| 378.108 | |
| JBDGDEWWOUBZPM-UHFFFAOYSA-N | |
| 2132 | |
| C1CC(CCC1NCC2=CC(=CC(=C2N)Br)Br)O |
| C13H18Br2N2O | |
| MFCD00078932 | |
| ambroxol, ambroxolum, bisolvon metabolite viii, bromhexine-metabolite viii, 4-2-amino-3,5-dibromobenzyl amino cyclohexanol, ambroxol inn:ban, ambroxol ep impurity d, ambroxol base, ambroxolum inn-latin, rac-cis-ambroxol | |
| 4-[(2-amino-3,5-dibromophenyl)methylamino]cyclohexan-1-ol |
Specifications
| 23828-92-4 | |
| C13H18Br2N2O | |
| 1 g | |
| ambroxol, ambroxolum, bisolvon metabolite viii, bromhexine-metabolite viii, 4-2-amino-3,5-dibromobenzyl amino cyclohexanol, ambroxol inn:ban, ambroxol ep impurity d, ambroxol base, ambroxolum inn-latin, rac-cis-ambroxol | |
| JBDGDEWWOUBZPM-UHFFFAOYSA-N | |
| 4-[(2-amino-3,5-dibromophenyl)methylamino]cyclohexan-1-ol | |
| 2132 | |
| 99% | |
| Ambroxol hydrochloride |
| White | |
| MFCD00078932 | |
| 14,386 | |
| Slightly soluble in water (∽5mg/ml) or ethanol (∽5mg/ml). | |
| C1CC(CCC1NCC2=CC(=CC(=C2N)Br)Br)O | |
| 378.108 | |
| 414.57 | |
| Powder |
Safety and Handling
EINECSNumber : 242-899-2
RTECSNumber : GV8423000
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only