missing translation for 'onlineSavingsMsg'
Learn More

Ambroxol hydrochloride, 98%

Catalog No. aaj6371203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6371203 1 g
AAJ6371206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6371203 Supplier Thermo Scientific Chemicals Supplier No. J6371203
Only null left

CAS: 23828-92-4 | C13H18Br2N2O | 378.108 g/mol

Ambroxol hydrochloride is a medication indicated to alleviate chest congestion associated with conditions that include bronchitis, pneumonia, bronchospasm asthma, cough, and allergy. Preclinically, ambroxol, the active ingredient of Mucosolvan, has been shown to increase respiratory tract secretion. It enhances pulmonary surfactant production and stimulates ciliary activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ambroxol hydrochloride is a medication indicated to alleviate chest congestion associated with conditions that include bronchitis, pneumonia, bronchospasm asthma, cough, and allergy. Preclinically, ambroxol, the active ingredient of Mucosolvan, has been shown to increase respiratory tract secretion. It enhances pulmonary surfactant production and stimulates ciliary activity.

Solubility
Slightly soluble in water (∼5 mg/ml) or ethanol (∼5 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 23828-92-4
Molecular Formula C13H18Br2N2O
Molecular Weight (g/mol) 378.108
MDL Number MFCD00078932
InChI Key JBDGDEWWOUBZPM-UHFFFAOYSA-N
Synonym ambroxol, ambroxolum, bisolvon metabolite viii, bromhexine-metabolite viii, 4-2-amino-3,5-dibromobenzyl amino cyclohexanol, ambroxol inn:ban, ambroxol ep impurity d, ambroxol base, ambroxolum inn-latin, rac-cis-ambroxol
PubChem CID 2132
IUPAC Name 4-[(2-amino-3,5-dibromophenyl)methylamino]cyclohexan-1-ol
SMILES C1CC(CCC1NCC2=CC(=CC(=C2N)Br)Br)O

Specifications

Color White
Quantity 1 g
Merck Index 14,386
Solubility Information Slightly soluble in water (∽5mg/ml) or ethanol (∽5mg/ml).
Formula Weight 414.57
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Ambroxol hydrochloride
EINECSNumber 242-899-2
RTECSNumber GV8423000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only