missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ AMBMP Hydrochloride, >98%

Catalog No. 604310
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
604310 10 mg
604350 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 604310 Supplier Tocris Bioscience™ Supplier No. 6043/10*
Only null left

Wnt canonical signaling activator; also tubulin polymerization inhibitor

Wnt canonical signaling activator. Also inhibits tubulin polymerization. Suppresses TLR2/4/5-induced inflammatory response in human monocytes. Inhibits cell proliferation and induces cell cycle arrest in MDA-MB-231 breast cancer cells. Also decreases embryo to blastocyst development.

Chemical Identifiers

CAS 2095432-75-8
Molecular Formula C19H19ClN4O3

Specifications

Quantity 10 mg
Solubility Soluble to 100mM in DMSO
Formula Weight 386.84
Purity >98%
Molecular Weight (g/mol) 386.836
Chemical Name or Material N4-(1,3-Benzodioxol-5-ylmethyl)-6-(3-methoxyphenyl)-2,4-pyrimidinediamine hydrochloride
Recommended Storage Store at -20°C