missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Yeast RNA (5 mg/mL)

Catalog No. AM7120G
Encompass
Change view
Click to view available options
Quantity:
1 x 0.5 mL Tube
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM7120G 1 x 0.5 mL Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM7120G Supplier Invitrogen™ Supplier No. AM7120G

Purified Saccharomyces cerevisiae yeast RNA is is a very effective blocking agent when used in Northern prehybridization and hybridization buffers, as well as being suitable as a coprecipitant in nucleic acid precipitations.

Purified Saccharomyces cerevisiae yeast RNA is suitable as a coprecipitant in nucleic acid precipitations. It cannot be used in reactions inhibited by exogenous RNA, and it is the most inexpensive source of a high-quality coprecipitant. This total RNA preparation is supplied in one tube containing 0.5 mL at a concentration of 5 mg/mL in diethyl pyrocarbonate-treated distilled water. It is typically used at a working concentration of 10–20 μg/mL.

What is a Coprecipitant?

Coprecipitants are inert substances used to aid recovery of nucleic acids during alcohol precipitations. While they can be used for precipitating large amounts of nucleic acids, they are essential for quantitative recovery of small amounts of nucleic acids in dilute solutions. Often, the use of such molecules is desirable for no other reason but visualization of the pelleted precipitate after centrifugation.

Order Info

Shipping Condition: Dry Ice

Specifications

Content And Storage Store at –20°C.
Concentration 5 mg/mL
For Use With (Application) Carrier for Nucleic Acid Precipitation
Product Type Yeast RNA
Quantity 1 x 0.5 mL Tube
Shipping Condition Dry Ice

For Research Use Only. Not for use in diagnostic procedures.