missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Yeast RNA (10 mg/mL)

Catalog No. AM7118
Encompass
Change view
Click to view available options
Quantity:
10 x 10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM7118 10 x 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM7118 Supplier Invitrogen™ Supplier No. AM7118

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Purified Torulla Invitrogen yeast RNA is suitable as a coprecipitant in nucleic acid precipitations. It cannot be used in reactions inhibited by exogenous RNA, and it is the most inexpensive source of a high-quality coprecipitant.

Purified Torulla Invitrogen yeast RNA is suitable as a coprecipitant in nucleic acid precipitations. It cannot be used in reactions inhibited by exogenous RNA, and it is the most inexpensive source of a high-quality coprecipitant. This total RNA preparation is supplied in ten 1 mL tubes at a concentration of 10 mg/mL in diethyl pyrocarbonate-treated distilled water. It is typically used at a working concentration of 10 to 20 μg/mL.

What is a Co-precipitant?
Coprecipitants are inert substances used to aid recovery of nucleic acids during alcohol precipitations. While they can be used for precipitating large amounts of nucleic acids, they are essential for quantitative recovery of small amounts of nucleic acids in dilute solutions. Often, the use of such molecules is desirable for no other reason but visualization of the pelleted precipitate after centrifugation.

Order Info

Shipping Condition: Dry Ice

Specifications

Content And Storage Store at –20°C.
Concentration 10 mg/mL
For Use With (Application) Carrier for Nucleic Acid Precipitation
Product Type Yeast RNA
Product Line Ambion
Quantity 10 x 10 mg
Shipping Condition Dry Ice

For Research Use Only. Not for use in diagnostic procedures.