missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ BSA (50 mg/mL)

Catalog No. AM2618
Encompass
Change view
Click to view available options
Quantity:
1 x 50 mg
5 x 50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM2618 5 x 50 mg
AM2616 1 x 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AM2618 Supplier Invitrogen™ Supplier No. AM2618

Ambion Ultrapure BSA is a 'non-acetylated' BSA pure enough to use when DNA or RNA integrity is critical.

Ambion Ultrapure BSA is a 'non-acetylated' BSA pure enough to use when DNA or RNA integrity is critical.

• Tubes containing 50 mg are provided, at a concentration of 50 mg/mL
• Bovine Serum Albumin (BSA) has many uses as a carrier protein and as a stabilizing agent in enzymatic reactions
• In northern, Southern, and dot blot hybridizations, BSA is also used as a blocking agent
• It is recommended in buffers for nick translation, polymerase reactions, and ligations

BSA is also a common additive for PCR amplifications, footprinting, and gel shift assays. In restriction digests, BSA has been shown to enhance enzyme activity. The molecular weight is 68 kD. Ultrapure BSA is rigorously tested for DNase, RNase, endonuclease, protease, peroxidase, and alkaline phosphatase activity, and assayed for fluorescence background. Exonuclease Activity: 3' - 5'.

Order Info

Shipping Condition: Dry Ice

Specifications

Content And Storage Store at –20°C.
Concentration 50 mg/mL
Product Line Ambion
Quantity 5 x 50 mg
Shipping Condition Dry Ice
Form Liquid
What is BSA and its function in PCR? What concentration of BSA do you recommend?

BSA, bovine serum albumin, is usually used in PCR as a stabilizer. A concentration of 160 µg/mL is typical. Gelatin may also be used as a stabilizer at 0.01% (w/v).

For Research Use Only. Not for use in diagnostic procedures.