Please login to your online account to display your discounted pricing

Invitrogen™ Ambion™ UltraPure BSA (50mg/mL) DFS Item


Excellent PCR stabilizer

Manufacturer: invitrogen™  LSAM2616

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. AM2616

  • / Each
 

Description & Specifications

Specifications

Quantity 1 x 50mg
Storage Requirements Store at -20°C.
Concentration 50mg/mL