Learn More
Invitrogen™ Yeast tRNA (10 mg/mL)
Invitrogen Yeast tRNA purified from brewer's yeast. When used at a final concentration of 10 to 20 μg/mL, yeast tRNA is an effective coprecipitant to aid in recovery of small amounts of nucleic acids.
Supplier: Invitrogen™ AM7119
Description
Invitrogen Yeast tRNA purified from brewer's yeast. When used at a final concentration of 10 to 20 μg/mL, yeast tRNA is an effective coprecipitant to aid in recovery of small amounts of nucleic acids. It is supplied in one tube containing 0.5 mL at a concentration of 10 mg/mL.
What is a Coprecipitant?
Coprecipitants are inert substances used to aid recovery of nucleic acids during alcohol precipitations. While they can be used for precipitating large amounts of nucleic acids, they are essential for quantitative recovery of small amounts of nucleic acids in dilute solutions. Often, the use of such molecules is desirable for no other reason but visualization of the pelleted precipitate after centrifugation.
Order Info
Shipping Condition: Dry Ice
Specifications
| Store at –20°C. | |
| Carrier for Nucleic Acid Precipitation | |
| Ambion | |
| Dry Ice |
| 10 mg/mL | |
| Yeast tRNA | |
| 0.5 mL |
For Research Use Only. Not for use in diagnostic procedures.