missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Tris (1 M), pH 7.0, RNase-free

Catalog No. am9851
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM9851 500 mL
AM9850G 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AM9851 Supplier Invitrogen™ Supplier No. AM9851
Only null left

Invitrogen Molecular biology grade, 1M Tris, pH 7.0 solution is supplied in one bottle. The buffer is certified RNase-free, economical, and ready-to-use.

Invitrogen Molecular biology grade, 1M Tris, pH 7.0 solution is supplied in one bottle. The buffer is certified RNase-free, economical, and ready-to-use. Due to the ubiquitous presence of RNases, manufacturing products for use with RNA is especially challenging. Invitrogen's nuclease-free reagents and buffers are manufactured in facilities specifically designed to prevent the introduction of nucleases. Highly sensitive RNase assays are performed at several different stages of the manufacturing process to ensure the highest quality. These reagents are rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.

Order Info

Shipping Condition: Room temperature

Specifications

Color Clear
Concentration 1 M
pH 7
Physical Form Liquid
Product Line Ambion
Quantity 500 mL
Formulation 1,3-Propanediol, 2-Amino-2-(Hydroxymethyl)-,Hydrochloride
Grade Molecular Biology

For Research Use Only. Not for use in diagnostic procedures.