missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TaqMan™ MicroRNA Cells-to-CT™ Kit
GREENER_CHOICE

Catalog No. 4391848
Encompass
Change view
Click to view available options
Quantity:
100 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4391848 100 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4391848 Supplier Invitrogen™ Supplier No. 4391848

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Includes cell lysis reagents, DNase, TaqMan MicroRNA RT reagents, and TaqMan Universal PCR Master Mix. 100 sample prep rxns, 100 DNase rxns, 100 reverse transcription rxns, and 500 20μL PCR rxns for 100 reactions.

First to offer a simple, complete workflow for miRNA expression analysis from a few samples, or it can easily be incorporated into automated, high throughput applications.

First to offer a simple, complete workflow for miRNA expression analysis from a few samples, or it can easily be incorporated into automated, high throughput applications.

  • Complete, validated solution: Optimized workflow includes cell lysis reagents, DNase, TAqMan MicroRNA RT reagents, and TAqMan Universal PCR Master Mix
  • Fast, simple, and convenient: Prepare samples in 10 minutes; eliminate tedious RNA isolation
  • Gold standard specificity: Achieve superior results with TAqMan MicroRNA reagents
  • Robust performance: Results equivalent to purified RNA
  • Broad input dynamic range: Linear detection from 10 to 100,000 cells
  • Fewer resources used, less hazardous, less waste

Order Info

Shipping Condition: Dry ice

Specifications

Content And Storage

Includes cell lysis reagents, DNase, TaqMan™ MicroRNA RT reagents, and TaqMan™ Universal PCR Master Mix.

• 100 sample prep reactions
• 100 DNase reactions
• 100 reverse transcription reactions,
• 500 x 20 μL PCR reactions

Detection Method Primer-probe
Format Tube
GC-Rich PCR Performance High
PCR Method 2-step RT-qPCR
Polymerase DNA Polymerase
Reaction Speed Standard
For Use With (Equipment) 7000 System, 7300 System, 7500 System, 7900HT System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, StepOne™, Standard Mode, StepOnePlus™, Standard Mode, QuantStudio 6 and 7 Pro systems, QuantStudio 7 Flex system, QuantStudio 3 and 5 systems, QuantStudio 12k Flex system, QuantStudio 6 Flex system
Passive Reference Dye ROX (Pre-mixed)
Product Line Ambion, Cells-to-CT, TaqMan
Product Type MicroRNA Cells-to-C Kit
Quantity 100 reactions
Reverse Transcriptase M-MLV
Shipping Condition Dry Ice
Sufficient For 100 Reactions
For Use With (Application) miRNA analysis
No. of Reactions 100 Reactions
Sample Type Cell
Show More Show Less
I'm seeing PCR products in the minus-RT control after performing my Cells-to-CT experiment. What does this mean?

If PCR products are seen in the minus-RT control reaction, but not in the no-template control, it indicates that genomic DNA remains in the sample and that genomic DNA was amplified in real-time PCR. Please follow the suggestions below:

- Ensure the DNase I is mixed thoroughly into the Lysis Solution.
- Use fewer cells per lysis reaction.
- Lyse cells using Lysis Solution that is at room temperature, and make sure that the lysis reaction occurs at room temperature.
You can also try increasing the incubation time of the lysis reaction to 8 minutes and/or using Lysis Solution that has been warmed up to 25 degrees C for cell lysis.

I'm getting PCR products in the no-template PCR control when performing a Cells-to-CT experiment. What could cause this?

PCR products in the no-template PCR control indicate that the sample is contaminated with DNA. More stringent steps need to be taken to control contamination.

I'm getting no PCR product or unexpected PCR products after performing a Cells-to-CT experiment. What could be the cause of this?

Please review the following possibilities and suggestions:

- A problem with adding or mixing the Stop Solution: ensure that the Stop Solution was added directly to the lysate, as components of the Lysis Solution may inhibit RT-PCR if not fully inactivated.
- The RNA was degraded: keep cells in PBS on ice before starting the cell lysis procedure.
- RNase in the sample was not completely inactivated: Too many cells could have been used or too much PBS left on the cells, diluting the lysis solution.
- The lysates sat too long before going to room temperature: Do not allow lysates to sit longer than 20 minutes at room temperature once the Stop Solution has been added.
- The sample does not contain the target RNA: Verify that the procedure is working by using the XenoRNA Control in the sample. Also check that your PCR primers can amplify your target under the PCR conditions you are using.

I ran out of stop solution for my Cells-to-CT experiment. Can I purchase it separately?

Yes, it is available in 1 mL aliquots (Cat. No. 4402960).

I have genomic DNA contamination in my Cells-to-CT reaction. How do I get rid of it?

1. Ensure that all medium is removed from the wells.
2. Wash with an equal volume of room temperature 1X PBS after the medium is removed.
3. Ensure that the reaction happens at room temperature (the lysis reaction may not reach room temperature if the plate is on ice, if the plate was quickly moved to the bench, or if a cold lysis solution was added).
4. Warm lysis solution to room temperature before adding to cells.
5. Allow the lysis reaction to proceed for 8 minutes at 25 degrees C.

Can I perform a Cells-to-CT experiment using virus infected cells?

While we have not tested this in house, the following paper did use our kit on cells infected with hepatitis C virus:
Triyatni M, Berger EA, Saunier B (2011) A new model to produce infectious hepatitis C virus without the replication requirement. PLoS Pathog 7(4)::e1001333.

What is the maximum amount of cDNA that can be used in real-time PCR for the TaqMan Gene Expression Cells-to-CT kit?

The TaqMan Gene Expression Cells-to-CT Kit can accommodate 45% of the total reverse transcriptase reaction volume as cell lysate.

What are the stopping points for the SYBR Cells-to-CT protocol?

Here are the potential stopping points:

- At the Cell Lysis step after addition of the Stop Solution and incubating for 2 minutes at room temperature. Do not allow lysates to remain at room temperature for longer than 20 minutes after adding the Stop Solution. Lysates can be stored on ice for a maximum of 2 hours, or at -20 degrees C or -80 degrees C for a maximum of 5 months.
- At the Reverse Transcription (RT) step after assembling the RT reaction. Once assembled, mix reactions gently, then centrifuge briefly to collect the contents at the bottom of the reaction vessel, and store at 4 degrees C for up to 4 hours.
- Completed RT reactions may be stored at -20 degrees C.

Are the reverse transcriptases in the TaqMan Cells-to-CT kits and SYBR Cells-to-CT kits the same?

No. However, the SYBR Cells-to-CT reverse transcriptase will work with the TaqMan mastermixes (but not the other way around).

Do you have data to demonstrate that SYBR Cells-to-CT kits perform with similar equivalence in purifying RNA?

Yes, both the Power SYBR Green Cells-to-CT Kit and the Fast SYBR Green Cells-to-CT Kit were compared to a traditional RNA purification method followed by real-time PCR analysis. Both kits show equivalent performance to results obtained using purified RNA over a broad set of gene targets. Read more about the SYBR Green Cells-to-CT kits here (https://www.thermofisher.com/us/en/home/references/ambion-tech-support/rna-isolation/tech-notes/using-sybr-green-for-real-time-rt-pcr.html).

Can Cells-to-CT kits be used with TaqMan Low Density Arrays?

Yes, Cells-to-CT kits can be used with TaqMan Low Density Arrays. Here is a poster showing the workflow with miRNAs: http://www.thermofisher.com/content/dam/LifeTech/global/life-sciences/DNARNAPurification/Files/0714/AACR%20C2CT%20miRNA%20Final.pdf.

How many times can I freeze and rethaw the Cells-to-CT kit reagents?

We have tested the reagents that are stored frozen with five to ten freeze-thaw cycles and have seen no effect on CT values. Up to five freeze-thaw cycles for the lysate samples have not had any significant effect on gene expression data.

Can I go directly from cells to real-time PCR without having to purify RNA?

Yes, we offer our Cells-to-CT kits that allow you to bypass RNA purification.

Can the the direct lysis approach to DNA/RNA analysis be used for bacterial RNA extraction?

Cells-to-CT kits were not designed for RNA analysis from bacteria. Some customers do lysozyme incubation before the Cells-to-CT reaction, but the RNA profile could change as a result of this.

Is the the direct lysis approach to DNA/RNA analysis applicable for extraction of viral RNA?

Some customers have isolated RNA from viruses. Unfortunately, the protein coat could cause problems. Another option would be to use the MagMax Viral Isolation Kit.

How do I assess RNA quality with the direct lysis approach to DNA/RNA analysis?

We don't normally monitor RNA quality for Cells-to-CT or any other lysis based methods. We rely on the qRT-PCR results. We have isolated RNA after the lysis reactions and seen that there is no RNA degradation.

Would the direct lysis approach to DNA/RNA analysis be a good alternative for very small samples?

With very small samples, it is always better to use a lysis-based solution so that you don't lose any of your sample. Cells-to-CT kits, for example, can accommodate as few as 10 cells. If your sample size is smaller than 10 cells, we recommend using the Single Cell-to-CT Kit.

With the the direct lysis approach to DNA/RNA analysis, how do I know if there is RT inhibition?

In order to monitor for RT inhibition, we recommend using the Cells-to-CT Control Kit (Cat. Nos. 4402959, 4386995). This kit includes an exogenous RNA that is not like any RNA in the databases. Adding this to the RT reaction allows you to monitor for inhibition.

Can I use the the direct lysis approach to DNA/RNA analysis for tissue samples?

Some customers have used Cells-to-CT kits for small samples, like drosophila eyeballs. Howeveer, Cells-to-CT kits were not designed for such applications.

With the direct lysis approach to DNA/RNA analysis, does the product simultaneously isolate DNA and RNA?

Cells-to-CT kits can be used to analyze both RNA and DNA. DNA is stabilized as well as the RNA. When doing the analysis, be sure to use a primer and probe set that crosses the exon:intron boundary when you are looking at the RNA.

With the direct lysis approach to DNA/RNA analysis, can I use heparin- or EDTA-treated blood?

Unforutnately, the Blood-to-CT and MagMAX kits cannot be used with blood from heparin- or EDTA-stabilized tubes. The reactions require the lysis reagent found in Tempus tubes. Also, the heparin- or EDTA-stabilized tubes don't stabilize the RNA profiles, so we recommend using Tempus tubes.

Can the direct lysis approach to DNA/RNA analysis be automated?

Yes, Cells-to-CT kits are great for automation, including the screening of siRNAs or small molecules.

With the direct lysis approach to DNA/RNA analysis, can I use the direct lysis solution for uncultured primary cells or my other cell lines?

We use multiple cells lines in-house, including primary human hepatocytes, stem cells, and differentiated stem cells. When using a new cell line, we recommend that a pilot experiment with a dilution of cells from 10-100,000 cells looking for a loss in linearity of results. Please refer to the manual for how to perform this experiment.

With the direct lysis approach to DNA/RNA analysis, is it possible to freeze the lysates at -80 degrees C, and for how long?

Yes, they are stable at -20 or -80 degrees C for up to 5 months and can be freeze/thawed several times without loss in performance.

With the direct lysis approach to DNA/RNA analysis, can I use cells that are frozen or stabilized with RNAlater solution?

For the Cells-to-CT lysis reaction, you can use fresh cells, frozen cells, or cells that have been stabilized with RNAlater solution. You just want to ensure that cells were washed once with PBS before going into the lysis reaction.

Can I use 48-, 24-, or 6-well plates for direct cell lysis prior to qPCR?

The 50 µL Cells-to-CT lysis reaction can be done in any tube or any size plate. If you are using a 6- or 24- well plate, you may need to scale up the lysis reaction to cover the bottom of the plate to lyse all the cells. Also, do not go under the 50 µL reaction volume because the variability when pipetting the stop solution could cause variability with results.

Will the Cells-to-CT system work with my special cell line?

There is no reason why the Cells-to-CT system shouldn’t work with any cell line. However, due to differences in cell size and composition, the maximum number of cells per lysis reaction may be slightly different for different cell lines. We recommend testing for inhibition and optimal cell input by using the TaqMan Cells-to-CT and SYBR Green Cells-to-CT Control kits.

Is gDNA contamination an issue when using the Cells-to-CT kits?

Cells-to-CT technology features a unique method for lysing cultured cells while removing genomic DNA (gDNA) and preserving RNA integrity. Therefore, by following the lysis/DNase treatment steps in the protocol, gDNA contamination will not be an issue for qRT-PCR.

Can multiplex reactions be run with the Cells-to-CT kits?

Yes. Cells-to-CT kits are compatible with multiplex qRT-PCR assays. If a large number of targets are being tested, we recommend combining it with TaqMan assays.

Have the Cells-to-CT kits been used with plant tissue?

We have not tested plant tissue internally. When working with plants, we suggest employing one of the following standard homogenization methods: (1) motorized homogenizer, (2) stainless steel bead beating, or (3) liquid nitrogen-mortar/pestle.

When using the TaqMan MicroRNA Cells-to-CT Kit (Cat. No. 4391848) can I store the prepared cDNA?

Completed RT reactions may be stored at –20°C.

When using the TaqMan MicroRNA Cells-to-CT Kit (Cat. No. 4391848) can I store the cell lysates?

Cell lystes can be stored on ice for about 2 hours or at -20°C/-80°C for under 5 months. However, as a best practice, it is best to work with freshly prepared lysates.

Can I scale down the lysis reaction volume when working with the TaqMan MicroRNA Cells-to-CT Kit (Cat. No. 4391848)?

No, we do not recommend scaling down the reaction volume. Please use 50 µL at all times. Scaling down could lead to variable results from incomplete inactivation of reagents.

Do the 2-step Cells-to-CT kits contain RNase inhibitor?

Yes, the Stop Solution provided in the 2-step Cells-to-CT kits contains RNase inhibitor.

Can I use the TaqMan Gene Expression Cells-to-CT kit for multiplexing?

The TaqMan Gene Expression Cells-to-CT kit has been validated for duplexing. If you want to set up a multiplex real-time PCR reaction with 3 assays, we recommend using the TaqMan Fast Advanced Cells-to-CT kit (https://www.thermofisher.com/order/catalog/product/A35374).

I am using a Cells-to-CT kit and I see signal from the genomic DNA in my real-time PCR results. How do I get rid of the genomic DNA contamination?

To prevent signal from genomic DNA in the Cells-to-CT real-time PCR reaction, we recommend using a TaqMan assay or primer set that spans an exon-exon boundary, and adding DNase I to degrade genomic DNA during the lysis reaction. For optimal DNase activity in the lysis reaction, we recommend the following:
1. Ensure all media is removed from the cells.
2. Wash each well or cell pellet with an equal volume of room temperature 1X PBS.
3. Ensure the lysis reaction happens at room temperature. The lysis reaction may not reach room temperature if the plate is on ice prior to adding Lysis Solution, or cold Lysis Solution is added.
4. Warm the Lysis Solution to room temperature before adding to the cells.
5. Perform the lysis reaction at 25 degrees C for up to 8 minutes.

Can I DNase-treat my Cells-to-CT cell lysate after the Stop Solution has been added to the lysis reaction?

The Cells-to-CT Stop Solution prevents the DNase from being active, even if you add more. If you need to perform additional DNase treatment of the cell lysate sample after the Stop Solution is added, we recommend purifying the RNA from the cell lysate using traditional methods and DNase-treating the purified RNA.

I am using the TaqMan Gene Expression Cells-to-CT kit. Can I use the TaqMan Fast Advanced Master Mix in place of the TaqMan Gene Expression Master Mix to set up the qPCR reaction?

Yes, the TaqMan Fast Advanced Master Mix can be used in place of the TaqMan Gene Expression Master Mix when setting up the qPCR reaction for the TaqMan Gene Expression Cells-to-CT kit.

Can I use the Cells-to-CT kits with plasma samples?

We have not tested the compatibility of Cells-to-CT kits with plasma samples and would not recommend using this kit for plasma samples. Please refer to the following table to select an RNA purification kit based on your sample type: https://www.thermofisher.com/us/en/home/life-science/dna-rna-purification-analysis/rna-extraction/rna-types/total-rna-extraction.html.

What primers are used in the reverse transcription step for the Cells-to-CT kits?

The following 2-step Cells-to-CT kits use a mixture of oligo dT and random primers for the reverse transcription step:
• TaqMan Fast Advanced Cells-to-CT Kit
• TaqMan Gene Expression Cells-to-CT Kit
• SYBR Green Fast Advanced Cells-to-CT Kit
• Power SYBR Green Cells-to-CT Kit
• Fast SYBR Green Cells-to-CT Kit

The following 1-step Cells-to CT kits require gene-specific primers for the reverse transcription step, so you can either use your SYBR qPCR primers or TaqMan assay primers:
• Cells-to-CT 1-Step TaqMan Kit
• Cells-to-CT 1-Step Power SYBR Green Kit

Can I scale down the lysis reaction volume for the Cells-to-CT kits?

We do not recommend scaling down the lysis reaction volume for the Cells-to-CT kits. Reducing the lysis reaction volume below 50 µL can lead to incomplete inactivation of reagents and cause variability with results.

How do I remove gDNA from my Cells-to-CT reaction?

1. Ensure that all media is removed from the wells.
2. Wash with an equal volume of room temperature 1X PBS.
3. Ensure that the lysis reaction happens at room temperature (the lysis reaction may not reach room temperature if the plate is on ice, quickly moved to the bench, or cold lysis solution is added).
4. Warm the lysis solution to room temperature before adding to the cells.
5. Perform lysis reaction at 25 degrees C for up to 8 mins.

What is the optimal number of cells to use per reaction with Cells-to-CT kits?

To identify the maximum number of cells to use for each reaction, we recommend testing a range of cellular input amounts by setting up a serial dilution. Instructions for determining the best cell number input with Fast Advanced Cells-to-CT kits can be found in the User Bulletin: Cell Input Optimization for SYBR Green Fast Advanced Cells-to-CT Kit (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0017931_CellInputOptimization_SYBRGreenFastAdvCells-to-CT_UB.pdf) and the User Bulletin: Cell input optimization for TaqMan Fast Advanced Cells-to-CT Kit (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0017932_CellInputOptimization_TaqManFastAdvCells-to-CT_UB.pdf). For the non-Fast Advanced Cells-to-CT kits, instructions can be found in the Pilot Experiment section of the protocol for each kit.

Which cell lines have been tested to work with the Cells-to-CT system?

Here is a short list of cell lines that have been tested with the Cells-to-CT system: HeLa, HepG2, primary hepatocytes, SK-N-AS, SK-N-SH, U-87 MG, ME-180, A549, Jurkat, PC-12, PT-K75, NIH/3T3, Raji, HEK-293, COS-7, CHO-K1, NCI-H460, DU-145, K562, U-2 OS, Huh-7, Neuro 2A, and BJ. For additional information please visit the following page: https://www.thermofisher.com/us/en/home/life-science/dna-rna-purification-analysis/rna-extraction/rna-types/total-rna-extraction/cells-to-ct-kits/cells-to-ct-faq.html


For Research Use Only. Not for use in diagnostic procedures.