missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ T7-Oligo(dT)24V Promoter Primer, Anchored (100 ng/μl)

Catalog No. AM5712
Encompass
Change view
Click to view available options
Quantity:
40 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM5712 40 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM5712 Supplier Invitrogen™ Supplier No. AM5712

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

The primer is supplied in a single tube.

For use in reverse transcription reactions to append a T7 RNA polymerase promoter to cDNA fragments

Each Invitrogen primer is PAGE purified and performance tested in reverse transcription and subsequent in vitro transcription reactions. Promoter primers are anchored with a G, A, or C (V).

Gene Expression Analysis and Genotyping, In Vitro Transcription, PCR and Real-Time PCR, Reverse Transcription

Order Info

Shipping Condition: Dry Ice

Specifications

Product Type Primer
Content And Storage The primer is supplied in a single tube and should be stored at –20°C.
Shipping Condition Dry Ice
Product Line Ambion
Quantity 40 Reactions

For Research Use Only. Not for use in diagnostic procedures.