missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SSC (20X), RNase-free

Catalog No. AM9765
Encompass
Change view
Click to view available options
Quantity:
500 mL
1L
4 x 1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM9765 4 x 1 L
AM9770 500 mL
AM9763 1L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AM9765 Supplier Invitrogen™ Supplier No. AM9765
Only null left

Molecular biology grade, Invitrogen 20X SSC solution is supplied in one bottle containing 1 L. The buffer is certified RNase-free, economical, and ready-to-use.

Molecular biology grade, Invitrogen 20X SSC solution is supplied in one bottle containing 1 L. The buffer is certified RNase-free, economical, and ready-to-use. Due to the ubiquitous presence of RNases, manufacturing products for use with RNA is especially challenging. Invitrogen™'s nuclease-free reagents and buffers are manufactured in facilities specifically designed to prevent the introduction of nucleases. Highly sensitive RNase assays are performed at several different stages of the manufacturing process to ensure the highest quality. These reagents are rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.

Order Info

Shipping Condition: Room temperature

Specifications

Content And Storage Store at room temperature.
Concentration 20X
Packaging Type Bottle
Product Line Ambion
Quantity 4 x 1 L
Shipping Condition Room Temperature
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.