Learn More
Invitrogen™ Single Cell-to-CT™ qRT-PCR Kit

Description
The Ambion Single Cell-to-CT™ Kit contains a complete validated workflow for gene expression analysis for samples containing 1–10 cells. Each kit contains reagents for sample preparation, reverse transcription, pre-amplification, and qPCR that have been optimized together in a simple workflow that can be completed in only five steps.
Pre-optimized protocol ensures success and saves time
Each workflow begins with a simple 7-minute sample prep, where cells are effectively and reproducibly lysed with minimal processing into a solution that is compatible with downstream RT-PCR, without the need for purification. This is followed by the use of Superscript™ RT for reverse transcription, the TaqMan™ PreAmp Master Mix to amplify the cDNA, and TaqMan Gene Expression Master Mix for qPCR. All four of these steps have been developed to provide maximum sensitivity. The entire cell sample is maintained in the same well throughout the procedure so there is no sample loss or dilution of the precious limited material that could affect sensitivity.
Single cell sensitivity
Sensitivity of real-time PCR results can be affected by the efficiency of sample preparation, reverse transcription, or amplification. The Single Cell-to-CT Kit has addressed each of these potential problem areas to create a solution that enables reliable and robust gene expression analysis from single cells with maximal sensitivity. Detection of single cell equivalents using the Single Cell-to-CT Kit demonstrates appropriate linearity and sensitivity of qPCR results compared to a 100 cell sample control with excellent technical reproducibility. As expected, individual single cells exhibit greater variability due to inherent biological heterogeneity. In addition, through inclusion of a cDNA pre-amplification step, targets of interest are accurately amplified prior to real-time PCR. This critical step boosts signal in an unbiased manner, extending the analysis potential of limited samples. The accuracy and signal enhancement of a 96-gene panel using pre-amplification is shown.
Superior performance and reproducibility compared to alternative methods
Traditional sample preparation methods using organic solvents, glass fiber filters, or magnetic beads are not suitable for single-cell analysis due to the loss of sample through incomplete RNA precipitation, binding, and elution. Additionally, most single-cell homebrew methods which involve simple boiling to lyse cells lack the ability to inactivate endogenous RNases to stabilize gene expression profiles and can result in chemical cleavage of the RNA and inhibitor carryover, including gDNA contamination, which can affect all qRT-PCR applications. In contrast, the Single Cell-to-CT Kit shows superior sensitivity and reproducibility for single-cell analysis compared to traditional purification or homebrew boiling methods.
- Pre-optimized workflow for real-time RT-PCR from single cells
- Maximum sensitivity for single-cell analysis
- Superior performance and reproducibility compared to alternative methods
- Complete kit convenience and easy-to-follow protocol
- Suitable for use with 1-step and 2-step qRT-PCR
- Less hazardous, less waste, fewer resources used
Order Info
Shipping Condition: Dry ice
Specifications
Specifications
| Content And Storage | 400 μL Single Cell DNase I, 4.0 mL Single Cell Lysis Solution (store at 4°C), 400 μL Single Cell Stop Solution, 600 μL Single Cell SuperScript™ RT, 50 mL TaqMan™ Gene Expression MasterMix (store at 4°C), 2.1 mL Single Cell Pre-Amp Mix, 1.2 mL Single Cell VILO™ RT Mix, Store kit components at -20°C except for Single Cell Stop Solution and TaqMan™ Gene Expression MasterMix, which should be stored at 4°C. |
| Detection Method | TaqMan |
| PCR Method | 2-step RT-qPCR |
| Reaction Speed | 7 min. |
| For Use With (Equipment) | 7000 System, 7300 System, 7500 System, 7900HT Fast System, 7900HT System, GeneAmp 9700, GeneAmp™ 2400, StepOne™, Fast Mode, StepOne™, Standard Mode, StepOnePlus™, Fast Mode, StepOnePlus™, Standard Mode, Veriti Thermal Cycler |
| Product Line | Ambion, Cells-to-CT |
| Purification Time | 7 min. |
| Quantity | 400 reactions |
| Shipping Condition | Dry Ice |
| For Use With (Application) | RT-PCR |
| Show More |
Frequently Asked Questions (FAQs)
If you are targeting a low-abundance gene, you may have trouble getting Ct values in a good, reliable range (Ct > 32). To increase the sensitivity of the assay, you may want to consider the following:
- Increase the amount of RNA input into your reverse transcription reaction, if possible
- Increase the amount of cDNA in your qPCR reaction (20% by volume max)
- Try a different reverse transcription kit, such as our SuperScript VILO Master Mix, for the highest cDNA yield possible
- Consider trying a one-step or Cells-to-CT type workflow (depending on your sample type)
With very small samples, it is always better to use a lysis-based solution so that you don't lose any of your sample. Cells-to-CT kits, for example, can accommodate as few as 10 cells. If your sample size is smaller than 10 cells, we recommend using the Single Cell-to-CT Kit.
There is no reason why the Cells-to-CT system shouldn’t work with any cell line. However, due to differences in cell size and composition, the maximum number of cells per lysis reaction may be slightly different for different cell lines. We recommend testing for inhibition and optimal cell input by using the TaqMan Cells-to-CT and SYBR Green Cells-to-CT Control kits.
Samples can be placed at room temperature for up to 30 minutes following lysis, and for up to 2 hours after adding Stop Solution. Cell lysates, cDNA, and preamplification samples can all be frozen for future processing.
We do not recommend normalizing to an endogenous control due to the biological variation and transcriptional noise exhibited by single cells. Because of this variation, normalization can actually increase the spread of calculated expression levels in single cells.
For Research Use Only. Not for use in diagnostic procedures.