missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SDS, 10% Solution, RNase-free

Catalog No. AM9822
Encompass
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM9822 500 mL
AM9823 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AM9822 Supplier Invitrogen™ Supplier No. AM9822

Molecular biology grade, Invitrogen SDS, 10% Solution is supplied in one bottle containing 500 mL.

Molecular biology grade, Invitrogen SDS, 10% Solution is supplied in one bottle. The buffer is certified RNase-free, economical, and ready-to-use. SDS is used as a component of both hybridization and wash buffers for northern and Southern blots and has many other uses around the lab.

Order Info

Shipping Condition: Room temperature

Specifications

Content And Storage Store at room temperature.
Concentration 0.1
Packaging Type Bottle
Product Type SDS
Product Line Ambion
Quantity 500 mL
Shipping Condition Room Temperature
Form Liquid
What does SDS Solution (Cat. No. AM9820, AM9822, AM9823) contain?

Ambion molecular biology grade SDS Solution (Cat. No. AM9820, AM9822, AM9823) consists of Sodium Dodecyl Sulfate (SDS) and water. The solution is certified to be RNase-free and ready-to-use. AM9820: 20% SDS Solution, RNase-free, 250 mL
AM9822: 10% SDS Solution, RNase-free, 500 mL
AM9823: 10% SDS Solution, RNase-free, 1 L

For Research Use Only. Not for use in diagnostic procedures.