missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Salmon Sperm DNA, sheared (10 mg/mL)

Catalog No. AM9680
Encompass
Change view
Click to view available options
Quantity:
10 x 10 mg tubes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM9680 10 x 10 mg tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM9680 Supplier Invitrogen™ Supplier No. AM9680

Sheared Salmon Sperm DNA is a very effective blocking agent when used in Northern prehybridization and hybridization buffers at a concentration of 100 to 200 μg/mL.

Sheared Salmon Sperm DNA is a very effective blocking agent when used in Northern prehybridization and hybridization buffers at a concentration of 100 to 200 μg/mL.

Using this product
This sheared salmon sperm DNA has been treated with Proteinase K to remove any contaminating nucleases, followed by organic extraction with phenol:chloroform and ethanol precipitation. Salmon Sperm DNA is rigorously tested for RNase and DNase contamination and is suspended in nuclease-free water in a 1mL tube at a concentration of 10 mg/mL (1 mL per tube).

Order Info

Shipping Condition: Dry Ice

Specifications

Content And Storage Store pure solution at –20°C
Concentration 10 mg/mL
For Use With (Application) Block Non-Specific Hybridization
Product Type Salmon Sperm DNA
Product Line Ambion
Quantity 10 x 10 mg tubes
Shipping Condition Dry Ice
How many freeze thaw cycles do you recommend for Invitrogen, Salmon Sperm DNA, sheared (Cat. No. AM9680)?

We do not have specific guidance on the number of freeze thaws for Salmon Sperm DNA, but it can be frozen and thawed multiple times.

Do you have an alternative for Salmon Sperm DNA, sheared (10 mg/mL) (Cat. No. AM9680) to use as a spike-in for standard curve preparation for ssDNA?

You can use Yeast tRNA (10 mg/mL) (Cat. No. AM7119). When used at a final concentration of 10–20 µg/mL, yeast tRNA is an effective coprecipitant to aid in recovery of small amounts of nucleic acids.For more information, see the following link.

What is the molecular size of Salmon Sperm DNA (Cat. No. AM9680)?

The molecular size of the Salmon Sperm DNA (Cat. No. AM9680) is around 200-500 bp.

For Research Use Only. Not for use in diagnostic procedures.