missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNase T1 (1 U/μL)

Catalog No. AM2283
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM2283 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM2283 Supplier Invitrogen™ Supplier No. AM2283
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

RNase T1, 10X RNA Structure Buffer, 1X Alkaline Hydrolysis Buffer, Yeast RNA, Precipitation/Inactivation Buffer, and Gel Loading Buffer II

Invitrogen™ RNase T1, RNA-Grade cleaves 3' of single-stranded G residues and is tested for purity to ensure that no contaminating ribonuclease activities are present that could cleave at unanticipated sites.

Invitrogen™ RNase T1, RNA-Grade cleaves 3′ of single-stranded G residues and is tested for purity to ensure that no contaminating ribonuclease activities are present that could cleave at unanticipated sites. Supplied in one tube of 200μL (1U/μL). RNase T1 can be used to perform boundary experiments to define the minimal RNA sequence required for selectable activities such as protein binding or catalysis. In addition to applications for RNA structural analysis, RNA-Grade ribonucleases can be used to map protein binding sites on RNAs by comparing cleavage patterns in the presence and absence of an RNA binding protein.

  • 100 Units is the amount of enzyme that yields an increase in absorption at 260 nm of 0.01428 units/min. at room temperature using 60 μg/mL yeast total RNA as a substrate.
  • RNase A (Cat. No. AM2274) cleaves 3′ of single-stranded C and U residues. Combinations of the single- and double-stranded specific ribonucleases can provide rapid analysis of the physical structure of an interesting RNA.

Order Info

Shipping Condition: Dry ice

Specifications

Concentration 1 U/μL
Content And Storage RNase T1, 10X RNA Structure Buffer, 1X Alkaline Hydrolysis Buffer, Yeast RNA, Precipitation/Inactivation Buffer, and Gel Loading Buffer II

Store all components at –20°C.
Format Tube
Enzyme RNase
Fidelity (vs. Taq) 10X
For Use With (Application) Real Time PCR (qPCR)
No. of Reactions 200
Product Line Ambion
Product Type Rnase T1 PCR Kit
Quantity 200 μL
Shipping Condition Dry Ice
Source RNA
Form Solution
Show More Show Less
I need to digest my total RNA for my ribonuclease protection assay. What RNase should I use?

Total RNA can be digested using RNAse A or RNase T1. A mix of both can be used in ribonuclease protection assays.

For Research Use Only. Not for use in diagnostic procedures.