Learn More
Invitrogen™ Ambion™ RNase A, affinity purified, 1 mg/mL
Rigorously tested for contaminating nonspecific endonuclease, exonuclease, and protease activity
$174.00 - $532.00
Specifications
| Concentration | 1 mg/mL |
|---|---|
| Content And Storage | -20°C |
| Shipping Condition | Dry Ice |
| Enzyme | RNase |
| Product Type | RNase A |
Description
Ambion RNase A is endonuclease that specifically cleaves 3' of U and C residues. Affinity Purified RNase A is intended for critical applications when absence of DNase and other nonspecific nuclease activities is essential.
- Supplied in one tube containing 200μg (1mg/mL)
- Preparation contains RNase B, a carbohydrate isoform of RNase A
DNA & RNA Purification & Analysis, DNA Extraction, Maxiprep, Midiprep, Miniprep, Nuclease Protection Assays, Nucleic Acid Gel Electrophoresis & Blotting, Plasmid DNA Purification
Order Info
Shipping Conditions: Dry ice
Specifications
| 1 mg/mL | |
| Dry Ice | |
| RNase A |
| -20°C | |
| RNase | |
| Ambion |
For Research Use Only. Not for use in diagnostic procedures.