missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNA Fragmentation Reagents DFS Item

For the fragmentation of aRNA prior to hybridization with oligonucleotide arrays

Supplier:  Invitrogen™ AM8740

Catalog No. AM8740


Only null left
Explore available promotions

Description

Description

The protocol is simple, fast and can be completed in less than 20 min. Sufficient Invitrogen™ reagents are included for 200 rxns. Many protocols for using amplified RNA in microarray analysis recommend fragmenting the aRNA prior to hybridization with an oligonucleotide microarray. This fragmentation step improves hybridization kinetics with the arrayed oligonucleotides and can lead to enhanced signal. The protocol is simple. Mix the aRNA with the Invitrogen 10X RNA Fragmentation Reagent and heat to 70°C for 15 min. The reaction is then stopped by the addition of the Stop Solution and placed on ice until ready for hybridization with the microarray. The average size of the resulting fragments will be 60-200 nucleotides. Up to 10μg of RNA can be fragmented in a 10μL rxn.

Gene Expression Analysis and Genotyping, Microarray Analysis

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

RNA Fragmentation Reagent
10X Fragmentation Reagent and Stop Solution should be stored at room temperature.
Ambion
Liquid
10 X
Room Temperature
200 Reactions
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.