Learn More
Invitrogen™ Ambion™ Recombinant RNase A
Rigorously tested for contaminating nonspecific endonuclease, exonuclease, and protease activity
Supplier: Invitrogen™ AM2269
Description
For large batch sizes, use of RNase is still only effective technique applicable to remove host RNA. Other methods such as selective precipitation, gel filtration, chromatography, and endogenous E. coli RNase digestion have only been effective for small-scale or bench-scale purification. This product was designed specifically to minimize the risk of introducing pathogens into bioprocessing.
- Substantially free of host RNA
- For research applications
DNA & RNA Purification & Analysis, DNA Extraction, Maxiprep, Midiprep, Miniprep, Plasmid DNA Purification
Order Info
Shipping Conditions: Dry ice
Specifications
| -20°C | |
| RNase | |
| Recombinant RNase A |
| Dry Ice | |
| 200 μg | |
| Ambion |
For Research Use Only. Not for use in diagnostic procedures.