missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Proteinase K Powder

Catalog No. AM2542
Encompass
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM2542 100 mg
AM2544 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AM2542 Supplier Invitrogen™ Supplier No. AM2542

Proteinase K from the fungus Engyodontium album is a non-specific serine protease that is useful for general digestion of proteins.

Proteinase K from the fungus Engyodontium album is a non-specific serine protease that is useful for general digestion of proteins.

Proteinase K Remains Active:
• Over a wide pH range (optimal activity between 6.5 and 9.5)
• Under denaturing conditions (e.g. the presence of SDS or urea)
• In the presence of metal chelating agents (e.g. EDTA)
• At comparatively high temperatures (optimum digestion temperature is 65°C)

Applications: Removal of endogenous nucleases during the preparation of DNA and RNA; preparation of tissue sections for in situ hybridization.

Performance and Quality Testing: Endodeoxyribonuclease and exodeoxyribonuclease assays; liquid form tested for absence of RNase activity.

Unit Definition: One mAnson unit is described as that amount of enzyme that liberates 1 μmole of Folin-positive amino acid within one minute at 37°C using hemoglobin as a substrate.

Order Info

Shipping Condition: Dry Ice

Specifications

Host Species T. album
Packaging Bottle
Content And Storage Store dry powder in desiccant at –20°C.
Shipping Condition Dry Ice
Quantity 100 mg
Product Type Proteinase K
Product Line Ambion
How do I isolate DNA from single bacterial colonies before PCR?

Scrape a single colony from the surface of an agar plate and transfer to 12 µL of SCL solution [10 mM Tris-HCl (pH 7.5), 1 mM EDTA, 50 µg/mL proteinase K] and incubate for 15 min at 55 degrees C. Inactivate proteinase K for 15 min at 80 degrees C and add 20 µL of deionized water. Centrifuge at 12,000 x g for 3 min. Transfer the supernatant to a new tube.

What are the recommended conditions for proteinase K treatment when isolating RNA or DNA samples?

Concentration: Generally proteinase K is used in the concentration range of 50 to 500 µg/mL at 65 degrees C in the presence of SDS (0.5-1%).

Temperature optimum: 65 degrees C; 12X more active at 65 degrees C than at 25 degrees C.

pH: Proteinase K is stable over a wide pH range (4.0 to 12.5), with optimal activity at pH 6.5 to 9.5. It is most stable at pH 8.
Inactivation: Heat inactivate Proteinase K at 80 degrees C for 15 min. Phenol extraction is the recommended method to ensure complete inactivation.

Inhibited by: PMSF (0.1 to 1.0 mM of PMSF is usually sufficient for inhibition of Proteinase K)

Not inhibited by: Proteinase K is not inactivated by metal ions, chelating agents (e.g., EDTA), sulfhydryl reagents or by trypsin or chymotrypsin inhibitors. Activity can be stimulated by addition of denaturing agents (SDS and urea).

For Research Use Only. Not for use in diagnostic procedures.