missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Proteinase K for MagMAX™-96 DNA Multi-Sample Kit

Catalog No. 4489110
Encompass
Change view
Click to view available options
Quantity:
96 preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4489110 96 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4489110 Supplier Invitrogen™ Supplier No. 4489110
Get Up to $3,000 of qPCR Products with Purchase
View all promotions

Includes

1 tube

This is the Proteinase K found in the MagMAX™-96 DNA Multi-Sample Kit, made available separately for applications that require more proteinase K enzyme than is provided in the kit.

This is the Proteinase K found in the MagMAX™-96 DNA Multi-Sample Kit (Cat. No. 4413020, 4413021, 4413022), made available separately for applications that require more proteinase K enzyme than is provided in the kit.

Specifications

Content And Storage 1 tube

Store at -25°C to -15°C.
Final Product Type Genomic DNA
For Use With (Equipment) 7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, Ion PGM™ System, KingFisher™ Flex Magnetic Particle Processor, OpenArray™, MagMAX™ Express-96 Standard Magnetic Particle Processor, MagMAX™ Express-96 Deep Well Magnetic Particle Processor
For Use With (Application) Next-Generation Sequencing, Real-Time Quantitative PCR (qPCR)
High-throughput Compatibility High-throughput Compatible, Not High-throughput Compatible (Manual), Automated Protocols
Product Line MagMAX
Product Type DNA Multi-Sample Kit
Quantity 96 preps
Purity DNase-Free
Target Genomic DNA
Form Liquid
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.