missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PK Buffer for MagMAX™-96 DNA Multi-Sample Kit

Catalog No. 4489111
Encompass
Change view
Click to view available options
Quantity:
11 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4489111 11 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4489111 Supplier Invitrogen™ Supplier No. 4489111
Get Up to $3,000 of qPCR Products with Purchase
View all promotions

Includes

2 tubes

This is the PK Buffer found in the MagMAX-96 DNA Multi-Sample Kit (Cat. No. 4413020, 4413021, 4413022), made available separately for applications that require more PK Buffer than is provided in the kit. DNase-free.

This is the PK Buffer found in the MagMAX-96 DNA Multi-Sample Kit (Cat. No. 4413020, 4413021, 4413022), made available separately for applications that require more PK Buffer than is provided in the kit. DNase-free.

For use with: 7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, Ion Personal Genome Machine™ (PGM™) System, KingFisher™ Flex Magnetic Particle Processor, MagMAX™ Express-96 Deep Well Magnetic Particle Processor, MagMAX™ Express-96 Standard Magnetic Particle Processor, OpenArray™

Specifications

Content And Storage 1 bottle

Store at room temperature.
Format 96-well Plate
Isolation Technology Magnetic Bead
Sample Type Blood, Buccal DNA Collector™ Sample, Buccal Samples, Buccal Swab, Cells, Liquid Samples (e.g. Serum), Saliva, Tissue (Mammalian), Urine, Whole Blood (Mammalian)
Final Product Type Genomic DNA
For Use With (Equipment) 7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, Ion PGM™ System, KingFisher™ Flex Magnetic Particle Processor, OpenArray™, MagMAX™ Express-96 Standard Magnetic Particle Processor, MagMAX™ Express-96 Deep Well Magnetic Particle Processor
For Use With (Application) Next-Generation Sequencing, Real-Time Quantitative PCR (qPCR)
High-throughput Compatibility High-throughput Compatible, Not High-throughput Compatible (Manual), Automated Protocols
No. of Reactions 96 Preps
Product Line MagMAX
Product Type DNA Multi-Sample Kit
Quantity 11 mL
Purity DNase-Free
Target Genomic DNA
Test Time 1 hr.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.