missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PBS - Phosphate-Buffered Saline (10X) pH 7.4, RNase-free

Catalog No. p-4926912
Encompass
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM9624 500 mL
AM9625 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AM9624 Supplier Invitrogen™ Supplier No. AM9624
Only null left

PBS (phosphate buffered saline) is a pH-adjusted blend of ultrapure-grade phosphate buffers and saline solutions.

PBS (phosphate buffered saline) is a pH-adjusted blend of ultrapure-grade phosphate buffers and saline solutions which, when diluted to a 1X working concentration, contains 137 mM NaCl, 2.7 mM KCl, 8 mM Na2HPO4, and 2 mM KH2PO4. Each 10X PBS solution is ready to use upon dilution to the desired concentration. This molecular biology-grade PBS is certified RNase-free; it is rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.

Order Info

Shipping Condition: Room temperature

Specifications

Chemical Name or Material Phosphate Buffered Saline (PBS)
Concentration 10 X
pH 7.4
Physical Form Liquid
Product Line Ambion
Quantity 500 mL

For Research Use Only. Not for use in diagnostic procedures.