Learn More
Invitrogen™ NaCl (5 M), RNase-free
Description
Invitrogen Molecular biology grade, 5 M NaCl solution is supplied in one bottle. The solution is certified RNase-free, economical, and ready-to-use. Due to the ubiquitous presence of RNases, manufacturing products for use with RNA is especially challenging. Invitrogen's nuclease-free reagents and buffers are manufactured in facilities specifically designed to prevent the introduction of nucleases. Highly sensitive RNase assays are performed at several different stages of the manufacturing process to ensure the highest quality. These reagents are rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Chemical Name or Material | Sodium Chloride |
| Content And Storage | Store at room temperature. |
| Concentration | 5 M |
| Packaging Type | Bottle |
| Product Line | Ambion |
| Purity | Molecular Biology Grade |
| Quantity | 100 mL |
| Shipping Condition | Room Temperature |
| Form | Liquid |
Frequently Asked Questions (FAQs)
Add 10 to 20 mM NaCl to the sample to maintain proper ionic strength.
For Research Use Only. Not for use in diagnostic procedures.