missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NaCl (5 M), RNase-free

Invitrogen Molecular biology grade, 5 M NaCl solution is supplied in one bottle. The solution is certified RNase-free, economical, and ready-to-use.

Supplier:  Invitrogen™ AM9759

Encompass

 Afficher plus de versions de ce produit

Numéro de catalogue. AM9759


Il en reste null

Description

Description

Invitrogen Molecular biology grade, 5 M NaCl solution is supplied in one bottle. The solution is certified RNase-free, economical, and ready-to-use. Due to the ubiquitous presence of RNases, manufacturing products for use with RNA is especially challenging. Invitrogen's nuclease-free reagents and buffers are manufactured in facilities specifically designed to prevent the introduction of nucleases. Highly sensitive RNase assays are performed at several different stages of the manufacturing process to ensure the highest quality. These reagents are rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.

Order Info

Shipping Condition: Room temperature

Spécifications

Identifiants chimiques

Store at room temperature.
5 M
Ambion
Room Temperature

Spécifications

Store at room temperature.
Liquid
Ambion
Room Temperature
5 M
Sodium Chloride
500 mL
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.