missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MgCl2 (1 M)

Catalog No. am9530g
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM9530G 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM9530G Supplier Invitrogen™ Supplier No. AM9530G

Molecular biology grade, Invitrogen 1M MgCl2 solution is supplied in one bottle containing 100 mL. The solution is certified RNase-free, economical, and ready-to-use.

Molecular biology grade, Invitrogen 1M MgCl2 solution is supplied in one bottle containing 100 mL. The solution is certified RNase-free, economical, and ready-to-use. Due to the ubiquitous presence of RNases, manufacturing products for use with RNA is especially challenging. Invitrogen's nuclease-free reagents and buffers are manufactured in facilities specifically designed to prevent the introduction of nucleases. Highly sensitive RNase assays are performed at several different stages of the manufacturing process to ensure the highest quality. These reagents are rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.

Order Info

Shipping Condition: Room temperature

Specifications

Chemical Name or Material Magnesium Chloride
Content And Storage Store at room temperature.
Concentration 1 M
For Use With (Application) RNA Applications
Product Line Ambion
Purity Molecular Biology Grade
Quantity 100 mL
Shipping Condition Room Temperature
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.