missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MagMAX™ Wash Solution 1 Concentrate

Catalog No. AM8504
Encompass
Change view
Click to view available options
Quantity:
205 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM8504 205 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM8504 Supplier Invitrogen™ Supplier No. AM8504

Invitrogen MagMAX Wash Solution 1 Concentrate is a component of all the MagMAX Isolation Kits that may be purchased separately.

Invitrogen MagMAX Wash Solution 1 Concentrate is a component of all the MagMAX Isolation Kits that may be purchased separately. Supplied in one 205 mL bottle.

Order Info

Shipping Condition: Room temperature

Specifications

Buffer Wash Buffer
Content And Storage Store MagMAX™ Wash Solution 1 Concentrate at 4°C or room temperature.
Product Line Ambion, MagMAX
Quantity 205 mL
Shipping Condition Room Temperature
How should I prepare MagMAX wash solutions 1 and 2 from standalone Wash Solution 1 Concentrate (Cat. No. AM8504) and Wash Solution 2 Concentrate (Cat. No. AM8640)?

For MagMAX Wash Solution 1 Concentrate (Cat. No. AM8504), add 70 mL isopropanol to 135 mL of wash buffer concentrate. This can be added directly to the AM8504 bottle for a total volume of 205 mL of wash solution 1. For MagMAX Wash Solution 2 Concentrate (Cat. No. AM8640), add 160 mL ethanol to 40 mL of wash buffer concentrate. This can be added directly to the AM8640 bottle for a total volume of 200 mL of wash solution 2.

For Research Use Only. Not for use in diagnostic procedures.