missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Linear Acrylamide (5 mg/ml) (1 ml Tube)

Catalog No. AM9520
Encompass
Change view
Click to view available options
Quantity:
5 x 1mL tubes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM9520 5 x 1mL tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM9520 Supplier Invitrogen™ Supplier No. AM9520

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Invitrogen Linear Acrylamide (5 mg/mL) is for use as a nucleic acid coprecipitant and is ideal for PCR and RT-PCR reactions since small amounts of contaminating nucleic acids present in other carriers could be amplified.

Invitrogen Linear Acrylamide (5 mg/mL) is for use as a nucleic acid coprecipitant and is ideal for PCR and RT-PCR reactions since small amounts of contaminating nucleic acids present in other carriers could be amplified.

  • Ideal for RT-PCR
  • Increases pellet mass
  • Quantitative recovery of low concentrations (ng/mL) of nucleic acid
  • Prevents pellet loss in nuclease protection assays

What is a Coprecipitant?
Co-precipitants are inert substances used to aid recovery of nucleic acids during alcohol precipitations. While they can be used for precipitating large amounts of nucleic acids, they are essential for quantitative recovery of small amounts of nucleic acids in dilute solutions. Often, the use of such molecules is desirable for no other reason but visualization of the pelleted precipitate after centrifugation. Linear acrylamide offers advantages over other coprecipitants in that it is chemically synthesized and not treated with reagents (e.g., proteases) derived from biological sources. Therefore, it is not contaminated with biological material making it ideal for use upstream of RT-PCR. It has been shown to precipitate pictogram amounts of DNA fragments larger than 20 base pairs while failing to precipitate shorter fragments and free nucleotides. Linear acrylamide is useful for separating reaction products from unincorporated nucleotides and from most oligonucleotide primers. It should be used at a final working concentration of 10 to 20 μg/mL. Linear acrylamide will not interfere with A260/280 readings. Supplied in 5 x 1 mL tubes.

Order Info

Shipping Condition: Dry Ice

Specifications

Chemical Name or Material Acrylamide
Content And Storage Store at –20°C.
Concentration 5 mg/mL
For Use With (Application) RNA Extraction
Product Line Ambion
Purity Molecular Biology Grade
Quantity 5 x 1mL tubes
Shipping Condition Dry Ice
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.