missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IPTG (Dioxane-Free)

Catalog No. AM9464
Encompass
Change view
Click to view available options
Quantity:
2 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM9464 5 g
AM9462 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AM9464 Supplier Invitrogen™ Supplier No. AM9464

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Invitrogen IPTG (Isopropyl-beta-D-thiogalactoside) is an inducer of the lac operon and is frequently used in conjunction with X-GAL to determine the lac phenotype of recombinant plasmids or phage.

Invitrogen IPTG (Isopropyl-beta-D-thiogalactoside) is an inducer of the lac operon and is frequently used in conjunction with X-GAL to determine the lac phenotype of recombinant plasmids or phage. IPTG is also used to induce the expression of cloned genes that are under control of the lac operon.

Order Info

Shipping Condition: Room temperature

Specifications

Chemical Name or Material IPTG (Isopropyl β-D-thiogalactopyranoside)
Content And Storage Store at –20°C.
Packaging Type Bottle
Product Line Ambion
Quantity 5 g
Shipping Condition Room Temperature
Form Powder

For Research Use Only. Not for use in diagnostic procedures.