missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Guanidine Thiocyanate

Catalog No. AM9422
Encompass
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM9422 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM9422 Supplier Invitrogen™ Supplier No. AM9422

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Guanidine Thiocyanate is an ultrapure, molecular biology grade reagent. It is free of detectable nuclease and protease activity and is a strong protein denaturant, as both the guanidinium cation and the thiocyanate anion are chaotropic agents.

Guanidine Thiocyanate is an ultrapure, molecular biology grade reagent. It is free of detectable nuclease and protease activity and is a strong protein denaturant, as both the guanidinium cation and the thiocyanate anion are chaotropic agents. It is provided in one bottle containing 500 g. It is recommended for isolation of RNA, especially for tissues such as pancreas with high levels of RNase activity. In solution, it is known as guanidinium thiocyanate. Guanidine Thiocyanate is thoroughly tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.

Order Info

Shipping Condition: Room temperature

Specifications

Content And Storage Store at room temperature.
For Use With (Application) RNA Extraction
Product Type Guanidine Isothiocyanate
Product Line Ambion
Purity >99%
Quantity 500 g
Shipping Condition Room Temperature
Form Powder
What salts can be used to precipitate total RNA? What are the advantages of each? Do they require ethanol?

The three salts that can be used are:

- Guanidine thiocyanate, which requires ethanol. Guanidine thiocyanate is a common agent used for isolating RNA and we recommend it especially for tissues high in ribonuclease activity, such as the pancreas or spleen.
- Ammonium acetate, which requires ethanol. Ammonium acetate is useful when reducing coprecipitation of unwanted dNTPs and oligosaccharides. However, it should not be used when the nucleic acid will be phosphorylated using T4 polynucleotide kinase, since this enzyme is inhibited by ammonium ions.
- Lithium chloride, which does not require ethanol. LiCl is very effective in precipitating RNA but does not efficiently precipitate protein or DNA. It also does not precipitate unincorporated nucleotides.

For Research Use Only. Not for use in diagnostic procedures.