Learn More
Invitrogen™ GTP Soln. (10 mM)
This Invitrogen ribonucleotide triphosphate (rNTP) solution is a component of the MAXIscript™ Kits and is supplied in one tube containing 100 μL.
Supplier: Invitrogen™ AM8130G
Description
This Invitrogen ribonucleotide triphosphate (rNTP) solution is a component of the MAXIscript™ Kits (Cat. No. AM1312, AM1316, AM1314, AM1326, AM1324, AM1318, AM1322, AM1320, AM1310, and AM1308) and is supplied in one tube containing 100 μL. Each is subjected to RNase testing as well as a functional assay to ensure high levels of incorporation.
Order Info
Shipping Condition: Dry Ice
Specifications
| Store at -20°C. | |
| In Vitro transcription | |
| Ambion | |
| Dry Ice |
| 10 mM | |
| Unlabeled | |
| 100 μL |
For Research Use Only. Not for use in diagnostic procedures.