missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EDTA (0.1 mM), pH 8.0, RNase-free

Catalog No. AM9912
Encompass
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM9912 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM9912 Supplier Invitrogen™ Supplier No. AM9912

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Invitrogen 0.1 mM EDTA is a commonly recommended storage solution for RNA samples. 0.1 mM EDTA is in nuclease-free, ultrapure water. Supplied in one bottle of 50 mL.

Invitrogen 0.1 mM EDTA is a commonly recommended storage solution for RNA samples. 0.1 mM EDTA is in nuclease-free, ultrapure water. Supplied in one bottle of 50 mL. The last step in every RNA isolation protocol is to resuspend the purified RNA pellet. After painstakingly isolating the RNA, it is crucial that the pellet be suspended and stored in a safe, RNase-free solution. 0.1mM EDTA is prepared from the purest reagents available, under scrupulously monitored conditions in a state-of-the-art manufacturing facility. It is then subjected to rigorous quality control procedures to guarantee their purity and performance.

Order Info

Shipping Condition: Room temperature

Specifications

Chemical Name or Material EDTA
Content And Storage Store at room temperature.
Concentration 0.1 M
Sample Type RNA
For Use With (Application) Nucleic Acid Gel Electrophoresis & Blotting
Packaging Type Bottle
pH 8
Product Line Ambion
Purity DNase-Free, RNase-Free
Quantity 50 mL
Shipping Condition Room Temperature
Form Liquid
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.