missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DNase I Buffer (10X)

Catalog No. AM8170G
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM8170G 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM8170G Supplier Invitrogen™ Supplier No. AM8170G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

An Invitrogen component of the MAXIscript™ Kits, the buffer is supplied in one tube containing 1 mL.

An Invitrogen component of the MAXIscript™ Kits (Cat. No. AM1312, AM1316, AM1314, AM1326, AM1324, AM1318, AM1322, AM1320, AM1310, and AM1308), the buffer is supplied in one tube containing 1 mL.

Order Info

Shipping Condition: Room temperature

Specifications

Content And Storage Store at –20°C.
Concentration 10X
For Use With (Application) Reverse Transcription
Product Line Ambion
Quantity 1 mL
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.