missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DNAclear™ Purification Kit

Catalog No. AM1756
Encompass
Change view
Click to view available options
Quantity:
20 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM1756 20 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM1756 Supplier Invitrogen™ Supplier No. AM1756
Only null left

Designed to purify double-stranded DNA (dsDNA) generated by enzymatic reactions such as cDNA synthesis and PCR

The DNAclear™ cDNA Purification Kit process is simple and fast. DNA amounts ranging from 1ng to 3μg can be efficiently purified and concentrated. The DNAclear procedure consists of three steps: 1) DNA is bound to the Microfilter Cartridge in the presence of the DNA Binding Buffer, 2) the DNA is washed with the cDNA Wash Buffer, and 3) the DNA is eluted with preheated nuclease-free water. The DNAclear Kit can be used to remove free nucleotides, oligonucleotides, proteins, and salts from dsDNA samples. The DNA that is recovered from the Microfilter Cartridge is ready for any application that requires highly pure DNA including in vitro transcription reactions.

  • Includes sufficient reagents for 20 purifications
  • Efficiently purify and concentrate dsDNA ranging from 1ng to 3μg
  • Ideal for downstream applications such as in vitro transcription
  • Simple and fast protocol: about 10 minutes

DNA and RNA Purification and Analysis, DNA Extraction, PCR Product Clean-Up

Order Info

Shipping Condition: Room Temperature

Specifications

Product Type cDNA Purification Kit
For Use With (Equipment) Centrifuge
Content And Storage cDNA Binding Buffer, cDNA Wash Buffer, Microfilter Cartridges + Collection Tubes, and Elution Tubes should be stored at room temperature. Nuclease-free water may be stored at any temperature.
Final Product Type DNA
High-throughput Compatibility Not High-throughput Compatible (Manual)
Sample Type PCR Products
Shipping Condition Room Temperature
Format Microtube
Product Line Ambion, DNAclear
For Use With (Application) PCR, Sequencing, In Vitro Transcription, Cloning
Quantity 20 Preps
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.