missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DNA Binding Beads for MagMAX™-96 DNA Multi-Sample Kit

Catalog No. 4489112
Encompass
Change view
Click to view available options
Quantity:
96 preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4489112 96 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4489112 Supplier Invitrogen™ Supplier No. 4489112
Get Up to $3,000 of qPCR Products with Purchase
View all promotions

Includes

1 tube

These are the DNA Binding Beads found in the MagMAX-96 DNA Multi-Sample Kit, made available separately for applications that require more DNA Binding Beads than are provided in the kit. DNase-free.

These are the DNA Binding Beads found in the MagMAX-96 DNA Multi-Sample Kit (Cat. No. 4413020, 4413021, 4413022), made available separately for applications that require more DNA Binding Beads than are provided in the kit. DNase-free.

For use with: 7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, Ion Personal Genome Machine™ (PGM™) System, KingFisher™ Flex Magnetic Particle Processor, MagMAX™ Express-96 Deep Well Magnetic Particle Processor, MagMAX™ Express-96 Standard Magnetic Particle Processor, OpenArray™

Specifications

Content And Storage 1 tube, store at 2–8°C, DO NOT FREEZE.
Format 96-well Plate
Isolation Technology Magnetic Bead
Sample Type Blood, Buccal DNA Collector™ Sample, Buccal Samples, Buccal Swab, Cells, Liquid Samples (e.g. Serum), Saliva, Tissue (Mammalian), Urine, Whole Blood (Mammalian)
Final Product Type Genomic DNA
For Use With (Equipment) 7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, Ion PGM™ System, KingFisher™ Flex Magnetic Particle Processor, OpenArray™, MagMAX™ Express-96 Standard Magnetic Particle Processor, MagMAX™ Express-96 Deep Well Magnetic Particle Processor
For Use With (Application) Next-Generation Sequencing, Real-Time Quantitative PCR (qPCR)
High-throughput Compatibility High-throughput Compatible, Not High-throughput Compatible (Manual), Automated Protocols
Product Line MagMAX
Product Type DNA Binding Bead
Quantity 96 preps
Purity DNase-Free
Target Genomic DNA
Test Time 1 hr.
Show More Show Less
Can I purchase the RNA Binding beads from the MagMAX MirVana Total RNA Isolation Kit (Cat. No. A27828) separately?

Yes, you can. The DNA Binding Beads for MagMAX-96 DNA Multi-Sample Kit (Cat. No. 4489112) are the same as that used in A27828.

For Research Use Only. Not for use in diagnostic procedures.